Learn More
Lysine Acetylated Polyclonal Antibody, Rockland

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401939
Description
Store at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid (dilute only prior to immediate use).
Lysine Acetylated Polyclonal antibody specifically detects Lysine Acetylated in Many samples. It is validated for Western BlotSpecifications
| Lysine Acetylated | |
| Polyclonal | |
| Unconjugated | |
| Acetyl Lysine; Acetylated Lysine; Acetyllysine | |
| Anti-Lysine Acetylated Antibody was prepared from whole rabbit serum produced by repeated immunizations with acetylated lysine conjugated KLH. | |
| 100 μg | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl, 50% glycerol and no preservative; pH 7.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| All species | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.