Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine Hydroxylase 2/PLOD2 Antibody (OTI6D1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272556AF594
Description
Lysine Hydroxylase 2/PLOD2 Monoclonal specifically detects Lysine Hydroxylase 2/PLOD2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Lysine Hydroxylase 2/PLOD2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PLOD2 | |
Recombinant protein fragment corresponding to amino acids 359-640 of human Lysine Hydroxylase 2/PLOD2 (NP_891988) produced in E.coli. | |
0.1 mL | |
5352.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6D1 | |
Western Blot | |
2-oxoglutarate 5-dioxygenase (lysine hydroxylase) 2, 2-oxoglutarate 5-dioxygenase 2, procollagen-lysine, telopeptide lysyl hydroxylase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction