Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Antibody (1E12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP151678SS
Description
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Monoclonal specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111 | |
| Mouse | |
| 147 kDa | |
| 0.025 mL | |
| Chromatin Modifiers, Epigenetics | |
| 55818 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1000000000000 | |
| Unconjugated | |
| Q9Y4C1 | |
| KDM3A | |
| Purified recombinant fragment of human JMJD1A expressed in E. coli. [UniProt# Q9Y4C1] | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction