Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Polyclonal specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A | |
Polyclonal | |
Rabbit | |
Chromatin Modifiers, Epigenetics | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
55818 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KECLPTKASSKAELEIANPPELQKHLEHAPSPSDVSNAPEVKAGVNSDSPNNCSGKKVEPSALACRSQNLKESSVKVDNESCCSRSNNKIQNAPSRKSVL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111 | |
KDM3A | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title