Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149602AF594
Description
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 1.14.11, EC 1.14.11.-, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A | |
Rabbit | |
Affinity Purified | |
Chromatin Modifiers, Epigenetics | |
9682.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
KDM4A | |
A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction