Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Rabbit anti-Human, Mouse, Rat, Clone: S01-7F2, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
| Antigen | Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A |
|---|---|
| Clone | S01-7F2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Monoclonal antibody specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 9682 | |
| IgG | |
| Affinity-purified |
| S01-7F2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 1.14.11, EC 1.14.11.-, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A | |
| A synthesized peptide derived from human Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A (Uniprot#: O75164) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title