Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody (OTI5B9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272553AF532
Description
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Monoclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
KDM4C | |
Full-length protein expressed in 293T cell transfected with human Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C expression vector. | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5B9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C | |
Mouse | |
0.1 mL | |
23081 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction