Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197310B
Description
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Polyclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q9UGL1 | |
KDM5B | |
A synthetic peptide made to an internal portion of the human JARID1B protein (between residues 800-850) [UniProt Q9UGL1] | |
0.1 mL | |
Cancer, Chromatin Modifiers, Epigenetics | |
10765 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction