Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$236.50 - $499.50
Specifications
| Antigen | Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 |
|---|---|
| Concentration | 1 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.Specifications
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
| KDM6B | |
| IgG | |
| Affinity Purified | |
| This antibody is specific for Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 and will not recognize other JMJD proteins. |
| 1 mg/mL | |
| Unconjugated | |
| RUO | |
| PBS | |
| 23135 | |
| JMJD3 antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human JMJD3. The immunogen is located within amino acids 1370 - 1420 of JMJD3. Amino Acid Squence: KVPGSRTPGHQENNNF | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 180 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title