Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $469.50
Specifications
Antigen | Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 |
---|---|
Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 to 5 μg/mL, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
PBS | |
23135 | |
IgG | |
Immunogen affinity purified |
Western Blot 1 to 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 to 5 μg/mL, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
Partial recombinant protein made to an internal portion of the human JMJD3 protein (amino acids 1520-1682) [UniProt O15054] | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title