Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106640MFV500
Description
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
Polyclonal | |
Flow Cytometry | |
EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
Synthetic peptide made to an internal portion of the mouse JMJD3 protein (within residues 600-650). [Swiss-Prot# Q5NCY0] | |
0.1 mL | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
23135 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction