Learn More
Lysine-Specific Demethylase 4C (JMJD2C) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Lysine-Specific Demethylase 4C (JMJD2C) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | KDM4C,GASC1,JHDM3C,JMJD2C,KIAA0780 |
| Host Species | Rabbit |
| Immunogen | KLH-Conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human Lysine-specific demethylase 4C. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.