Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lysis Buffer
SDP

Catalog No. NBP349170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349170 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349170 Supplier Novus Biologicals™ Supplier No. NBP349170
Only null left
Add to Cart
Add to Cart

For use with select ELISA Kits.

Specifications

Quantity 20 mL
Content And Storage Store at 4 C short term. Store (at -20 C long term. Avoid freeze-thaw cycles.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.