Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Lysis Matrix S, 50mL Tubes
Click to view available options
Quantity:
10 x 50 mL
50 x 50 mL
Description
- Each impact-resistant 2mL tube contains 3.175mm (0.125 in.) diameter stainless steel beads
- Use to break hard/brittle samples
- Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
Certifications
DNase and RNase free

Specifications
Specifications
Content And Storage | Room temperature (15° - 30°C) |
Sample Type | All Samples, Bone, Fixed Samples, Food, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Spores, Tough Tissues |
For Use With (Equipment) | FastPrep-24, FastPrep 120, FastPrep-96 |
For Use With (Application) | Sample preparation, Sample lysis, Nucleic acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis; Gene function, Gene regulation |
Matrix | S |
Product Type | Lysis Matrix |
Quantity | 10 x 50 mL |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction