Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYSMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555755 Supplier Invitrogen™ Supplier No. PA555755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTPIHDLSAS DFLKKLDSQI SLSKKAAAQK LKKGENGVPG EDAGLHLSSP WMQQRAVLGP VPLTRTSRTR TLRDQEDEIF KL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

LYSMD1 (LysM Domain Containing 1) is a Protein Coding gene. Diseases associated with LYSMD1 include 3-Methylglutaconic Aciduria, Type Vii. An important paralog of this gene is LYSMD2. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYSMD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LYSMD1
Gene Accession No. Q96S90
Gene Alias 2610022K04Rik; AI415311; lysM and putative peptidoglycan-binding domain-containing protein 1; LysM domain containing 1; LysM, putative peptidoglycan-binding, domain containing 1; Lysmd1; RP11-68I18.5; SB145
Gene Symbols LYSMD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYSMD1. Recombinant protein control fragment (Product #RP-94422).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.