Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYSMD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584647
Description
Immunogen sequence: PILTEPRDLF NGLDSEEEKD GEEKVHPSNS EVWPHSTERK KQETGAGRAN GEVLPTPGQE T.
LYSMD1 (LysM Domain Containing 1) is a Protein Coding gene. Diseases associated with LYSMD1 include 3-Methylglutaconic Aciduria, Type Vii. An important paralog of this gene is LYSMD2. [GeneCards].
Specifications
LYSMD1 | |
Polyclonal | |
Unconjugated | |
LYSMD1 | |
2610022K04Rik; AI415311; lysM and putative peptidoglycan-binding domain-containing protein 1; LysM domain containing 1; LysM, putative peptidoglycan-binding, domain containing 1; Lysmd1; RP11-68I18.5; SB145 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96S90 | |
LYSMD1 | |
Recombinant protein corresponding to Human LYSMD1. Recombinant protein control fragment (Product #RP-107341). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction