Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYSMD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570125 Supplier Invitrogen™ Supplier No. PA570125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Zebrafish: 79%.

LYSMD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYSMD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LYSMD2
Gene Accession No. Q8IV50
Gene Alias 2210402C18Rik; AV033872; AW538442; LysM and putative peptidoglycan-binding domain-containing protein 2; LysM domain containing 2; LysM, putative peptidoglycan-binding, domain containing 2; LYSMD2; si:ch211-12a1.4; splice variant LysMD2B; zgc:91941
Gene Symbols LYSMD2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LYSMD2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256586
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.