Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYSMD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559188
Description
Immunogen sequence: LNIPVISEKP LLFNGLNSID SPENETADNS FSQEEEPVVA GEDLPPPSPQ ESDVQPVQPE EVSARDFLQR LDLQIKLSTQ AAKKLKEESR DEESPYATSL Y Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.
LYSMD2 is a protein coding gene.
Specifications
LYSMD2 | |
Polyclonal | |
Unconjugated | |
LYSMD2 | |
2210402C18Rik; AV033872; AW538442; LysM and putative peptidoglycan-binding domain-containing protein 2; LysM domain containing 2; LysM, putative peptidoglycan-binding, domain containing 2; LYSMD2; si:ch211-12a1.4; splice variant LysMD2B; zgc:91941 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
256586 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV50 | |
LYSMD2 | |
Recombinant protein corresponding to Human LYSMD2. Recombinant protein control fragment (Product #RP-98091). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction