Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYSMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570969 Supplier Thermo Scientific Supplier No. PA570969
Add to Cart
Add to Cart

LYSMD3 Polyclonal Antibody

LYSMD3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LYSMD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LYSMD3
Gene Accession No. Q7Z3D4
Gene Alias 1110030H10Rik; BC003322; lysM and putative peptidoglycan-binding domain-containing protein 3; LysM domain containing 3; LysM, putative peptidoglycan-binding, domain containing 3; LYSMD3
Gene Symbols LYSMD3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human LYSMD3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116068
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.