Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYSMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113060
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113060 Supplier Invitrogen™ Supplier No. PA5113060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYSMD3 Polyclonal Antibody for Western Blot, ELISA

LYSMD3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYSMD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LYSMD3
Gene Accession No. Q7Z3D4, Q99LE3
Gene Alias 1110030H10Rik; BC003322; lysM and putative peptidoglycan-binding domain-containing protein 3; LysM domain containing 3; LysM, putative peptidoglycan-binding, domain containing 3; LYSMD3
Gene Symbols LYSMD3
Host Species Rabbit
Immunogen Recombinant Human LysM and putative peptidoglycan-binding domain-containing protein 3 protein (1-127AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116068, 80289
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.