Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lysobacter enzymogenes Beta-lytic metalloendopeptidase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117605
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117605 Supplier Invitrogen™ Supplier No. PA5117605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Beta-lytic metalloendopeptidase is present in Achromobacter lyticus and Lysobacter enzymogenes. This enzyme catalyses the following chemical reaction: cleavage of N-acetylmuramoyl-/-Ala, and of the insulin B chain at 23-Gly-/-Phe-24 > 18-Val-/-Cys(SO(3)H).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysobacter enzymogenes Beta-lytic metalloendopeptidase
Applications Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias Beta-lytic protease
Host Species Rabbit
Immunogen Recombinant Lysobacter enzymogenes Beta-lytic metalloendopeptidase protein (1-178aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.