Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysozyme Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB6008500.1MG
Description
Lysozyme Polyclonal specifically detects Lysozyme in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Lysozyme | |
Polyclonal | |
HRP | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate | |
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis | |
Rabbit | |
Delipidation and Defibrination | |
RUO | |
4069 | |
Reconstitute with 0.1 ml deionized water (or equivalent) to desired concetration. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1:1000 - 1:5000, ELISA 1:10000, Immunohistochemistry 1:500 - 1:2500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500 - 1:2500 | |
P61626 | |
LYZ | |
This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length protein corresponding to amino acids 1-129 of Hen Egg White Lysozyme. | |
0.1 mg | |
Primary | |
This purified antibody is directed against lysozyme from hen egg white protein. | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction