Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysozyme Antibody (LZ598-10G9), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262223DL594
Description
Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Lysozyme | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LYZ | |
human lysozyme | |
0.1 mL | |
Primary | |
The antibody MEM-188 reacts with a 180 kDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that the antibody MEM-188 could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry | |
LZ598-10G9 | |
Flow Cytometry, Immunocytochemistry | |
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis | |
Mouse | |
Protein A purified | |
Cardiovascular Biology, Immunology | |
4069 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction