Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysozyme Antibody (OTI1C9), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272557IR
Description
Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lysozyme | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LYZ | |
Full length recombinant protein of human Lysozyme (NP_000230) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1C9 | |
Western Blot, Immunohistochemistry | |
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology, Immunology | |
4069.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction