Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysozyme Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2G5A8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Lysozyme |
|---|---|
| Clone | 2G5A8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6882
![]()
|
Proteintech Group Inc
664561IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Lysozyme is synthesized predomitly in reactive histiocytes rather than in resting, unstimulated phagocytes.Specifications
| Lysozyme | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 1,4 beta N acetylmuramidase C, lysozyme, lysozyme (renal amyloidosis), Lysozyme C, LYZ, LZM | |
| LYZ, Lyz1, Lyz2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2G5A8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| P17897, P61626, Q05820 | |
| 100157211, 17110, 25211, 4069 | |
| Lysozyme Fusion Protein Ag6917 | |
| Primary | |
| -20°C | |
| LYZ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title