Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme/Muramidase Ab-1 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB372A1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Ready-to-Use
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB372A1 500 μL Concentrated
RB372R1 1 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RB372A1 Supplier Epredia Supplier No. RB372A1

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Lysozyme/Muramidase Ab-1, Rabbit Polyclonal Antibody.

Lysozyme is synthesized predominantly in reactive histiocytes rather than in resting, unstimulated phagocytes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey, Pig, Cat and Dog. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human lysozyme purified from the urine of patients with monocytic leukemia

Positive Control: Tonsil, colon or skin

Cellular Localization: Cytoplasmic

Ab-1 was adsorbed with human plasma and urine proteins to remove the cross-reacting antibodies. Ab-1 labels myeloid cells, histiocytes, granulocytes, macrophages, and monocytes. This antibody is helpful in the identification of myeloid or monocytic nature of acute leukemia.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Lysozyme/Muramidase Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Human lysozyme purified from the urine of patients with monocytic leukemia
Quantity 500 μL
Regulatory Status IVD
Research Discipline Enzyme
Primary or Secondary Primary
Target Species Canine, Feline, Human, Non-human Primate
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.