Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lysozyme PROMO

Catalog No. PI89833PM
Click to view available options
:
Thermo Scientific™ Lysozyme

Catalog No. PI89833PM

Supplier: Thermo Scientific™ 89833

Degrade unwanted bacterial cell wall in protein or nucleic acid extracts

Lysozyme is an enzyme used to break down bacterial cell walls to improve protein or nucleic acid extraction efficiency. Lysozymes (muramidases) are a family of enzymes with antimicrobial activity characterized by the ability to damage the cell wall of bacteria. The enzyme acts by catalyzing the hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between the N-acetyl-D-glucosamine residues in chitodextrins. Although hen egg white lysozyme is most effective for the lysis of gram-positive bacteria, it also facilitates the lysis of gram-negative bacteria such as Salmonella and Shigella. The lysis of E. coli is especially improved by the addition of both lysozyme and a nucleases such as DNase I.

Highlights:

  • Bacterial cell wall lytic enzyme that improves inclusion body protein purification
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • Available in two convenient formats: lyophilized powder for large volume applications (Part No. 89833) and convenient glycerol freezer stock (Part No. 90082)

Properties of lysozyme:

  • Enzymatic function: Catalyzes the hydrolysis of 1,4-β-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between N-acetyl-D-glucosamine residues in chitodextrins
  • Molecular weight: 14.388 kDa
  • Extinction coefficient: ∽26.4 at 280nm
  • Isoelectric point: pH 11.0
  • Unit definition: 1 unit is that amount of enzyme needed to catalyze a decrease in absorbance at 450nm of 0.001/min at 25 degrees at pH 6.24 in a 1 cm cuvette due to lysis using a ∼0.25mg/mL suspension of Micrococcus luteus.

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Upon receipt, store at -20°C in a nonfrost-free freezer. Product shipped with ice.
Form Lyophilized
Reagent Type Enzyme for Cell Lysis
Enzyme Lysozyme (Muramidase)
Quantity 5 g

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.