Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody (OTI8B2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272558R
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Lysyl Oxidase Homolog 2/LOXL2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LOXL2 | |
Recombinant protein fragment corresponding to amino acids 152-450 of human Lysyl Oxidase Homolog 2/LOXL2 (NP_002309) produced in E.coli. | |
0.1 mL | |
4017.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI8B2 | |
Immunohistochemistry | |
EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction