Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100726AF532
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
LYVE-1 | |
Polyclonal | |
Western Blot | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
Rabbit | |
35 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
LYVE1 | |
This LYVE-1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human LYVE1 protein sequence (between residues 250-322). [UniProt# Q9Y5Y7] | |
Affinity Purified | |
Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
10894.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction