Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$219.50 - $524.00
Specifications
Antigen | LYVE-1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:500, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31776824), Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200, Flow (Intracellular) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100725SS
![]() |
Novus Biologicals
NB100725SS |
0.025 mL |
Each for $219.50
|
|
|||||
NB100725
![]() |
Novus Biologicals
NB100725 |
0.1 mL |
Each for $524.00
|
|
|||||
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
LYVE-1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:500, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31776824), Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200, Flow (Intracellular) | |
Polyclonal | |
Rabbit | |
Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
PBS with 0.02% Sodium Azide | |
10894 | |
This LYVE-1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the mouse LYVE1 protein sequence (between residues 250-318). [UniProt# Q8BHC0] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
LYVE1 | |
IgG | |
Affinity Purified | |
35 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title