Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100726V
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
LYVE-1 | |
Polyclonal | |
Western Blot | |
Q9Y5Y7 | |
LYVE1 | |
This LYVE-1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human LYVE1 protein sequence (between residues 250-322). [UniProt# Q9Y5Y7] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
Rabbit | |
35 kDa | |
0.1 mL | |
Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
10894 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction