Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYVE-1 Antibody, Novus Biologicals™
SDP

Catalog No. p-7200428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018198 0.025 mg
NB6001005 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018198 Supplier Novus Biologicals Supplier No. NB60010050.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYVE-1 Polyclonal specifically detects LYVE-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence, CyTOF-ready.

Specifications

Antigen LYVE-1
Applications CyTOF, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, Flow Cytometry 3-10 ug/mL, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 6-30 ug/mL, CyTOF-ready
Formulation 5mM PBS (pH 7.2) with No Preservative
Gene Alias cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1
Gene Symbols LYVE1
Host Species Rabbit
Immunogen Recombinant human LYVE-1
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10894
Test Specificity LYVE1
Reconstitution Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.