Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Antibody (RM0033-4D17) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$207.00
Specifications
Antigen | LYVE-1 |
---|---|
Clone | RM0033-4D17 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018003
![]() |
Novus Biologicals
NB110610260.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB11061026
![]() |
Novus Biologicals
NB11061026 |
0.1 mg | N/A | N/A | N/A | ||||
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LYVE-1 | |
LYOPH | |
Unconjugated | |
Rat | |
Q8BHC0 | |
10894 | |
IgG2 | |
Protein G purified | |
Mouse. Not yet tested in other species. |
RM0033-4D17 | |
Monoclonal | |
Purified | |
RUO | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
LYVE1 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title