Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Rabbit anti-Human, Mouse, Rat, Clone: 4G7C3, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316385100UL
Description
LYVE-1 Monoclonal antibody specifically detects LYVE-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
LYVE-1 | |
Monoclonal | |
Unconjugated | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1, UNQ230/PRO263, XLKD1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LYVE-1 (Q9Y5Y7). | |
100 μg | |
Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
10894 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
4G7C3 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction