Learn More
Invitrogen™ LYVE1 Polyclonal Antibody, Biotin

Description
Suggested positive control: mouse spleen lysate.
Specifications
Specifications
| Antigen | LYVE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.02 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Lyve1 |
| Gene Accession No. | Q8BHC0, Q9Y5Y7 |
| Gene Alias | 1200012G08Rik; cell surface retention sequence binding protein-1; cell surface retention sequence-binding protein 1; CRSBP1; CRSBP-1; extra cellular link domain-containing 1; extracellular link domain containing 1; extracellular link domain-containing 1; extracellular link domain-containing protein 1; HAR; hyaluronic acid receptor; lymphatic endothelial hyaluronan receptor LYVE-1; lymphatic vessel endothelial HA receptor-1; lymphatic vessel endothelial HA recptor-1; lymphatic vessel endothelial hyaluronan receptor 1; lymphatic vessel endothelial hyaluronic acid receptor 1; LYVE1; LYVE-1; sLyve 1; sLyve1; soluble LYVE 1; soluble LYVE1; UNQ230/PRO263; XLKD1 |
| Gene Symbols | Lyve1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.