Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYVE1 Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA522788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522788 Supplier Invitrogen™ Supplier No. PA522788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: mouse spleen lysate.

LYVE1 has been identified as a major receptor for HA (extracellular matrix glycosaminoglycan hyaluronan) on the lymph vessel wall. The deduced amino acid sequence of LYVE1 predicts a 322-residue type I integral membrane polypeptide 41% similar to the CD44 HA receptor with a 212-residue extracellular domain containing a single Link module, the prototypic HA binding domain of the Link protein superfamily. Like CD44, the LYVE1 molecule binds both soluble and immobilized HA. However, unlike CD44, the LYVE1 molecule colocalizes with HA on the luminal face of the lymph vessel wall and is completely absent from blood vessels. Hence, LYVE1 is the first lymph-specific HA receptor to be characterized and is a uniquely powerful marker for lymph vessels themselves. LYVE1 is a type I integral membrane glycoprotein. LYVE-1 is expressed primarily on lymphatic vessel endothelium and is likely to be involved in regulating the traffic of leucocytes and tumor cells to lymph nodes. The lymphatic vasculature forms a second circulatory system that drains extracellular fluid from the tissues and provides an exclusive environment in which immune cells can encounter and respond to foreign antigen. A number of molecules have been identified as markers for lymphatic endothelium which include LYVE1, PALE, VEGFR3, and podoplanin. Diseases associated with LYVE1 dysfunction includes Complete Androgen Insensitivity Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYVE1
Applications Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene Lyve1
Gene Accession No. Q8BHC0, Q9Y5Y7
Gene Alias 1200012G08Rik; cell surface retention sequence binding protein-1; cell surface retention sequence-binding protein 1; CRSBP1; CRSBP-1; extra cellular link domain-containing 1; extracellular link domain containing 1; extracellular link domain-containing 1; extracellular link domain-containing protein 1; HAR; hyaluronic acid receptor; lymphatic endothelial hyaluronan receptor LYVE-1; lymphatic vessel endothelial HA receptor-1; lymphatic vessel endothelial HA recptor-1; lymphatic vessel endothelial hyaluronan receptor 1; lymphatic vessel endothelial hyaluronic acid receptor 1; LYVE1; LYVE-1; sLyve 1; sLyve1; soluble LYVE 1; soluble LYVE1; UNQ230/PRO263; XLKD1
Gene Symbols Lyve1
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human LYVE1 protein sequence (between residues 250-322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10894, 114332
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.