Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYZL1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241184
Description
LYZL1 Polyclonal specifically detects LYZL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
LYZL1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
LYZL1 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human. The immunogen is located within the first 50 amino acids of LYZL1. Amino Acid Squence: SADSTEKSASGAGTRNL | |
Affinity Purified | |
RUO | |
Primary | |
At least two alternatively spliced transcript variants have been observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA | |
EC 3.2.1.17, KAAG648, LYC2bA534G20.1, lysozyme-like 1, lysozyme-like protein 1, MGC33408, PRO1278 | |
Rabbit | |
21 kDa | |
0.1 mg | |
Lipid and Metabolism | |
84569 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction