Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYZL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534443
Description
A suggested positive control is A-20 cell lysate. PA5-34443 can be used with blocking peptide PEP-1485.
The lysozyme family of proteins has an important role in host defense and is associated with the monocyte-macrophage system and enhances the activity of immunoagents. The mammalian lysozyme gene family is much larger than previously appreciated and consists of at least eight distinct genes scattered around the genome. C-type lysozymes are specifically involved catalyzing the hydrolysis of Beta-1,4 glycosidic bonds of the peptidoglycan of bacterial cell walls. As a homolog of human C-type lysozyme, LYZL1 is an intra-acrosomal, nonbacteriolytic, c lysozyme-like protein recently isolated from human spermatozoa.
Specifications
LYZL1 | |
Polyclonal | |
Unconjugated | |
Lyzl1 | |
1700038F02Rik; bA534G20.1; KAAG648; LYC2; lysozyme D1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; LYZD1; Lyzl1; PRO1278; RGD1559869; UNQ648/PRO1278 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
364745, 67328, 84569 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6UWQ5, Q9CPX3 | |
Lyzl1 | |
A 17 amino acid synthetic peptide near the amino terminus of human LYZL1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction