Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYZL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589463 Supplier Invitrogen™ Supplier No. PA589463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQDAPLSCLS PTKWSSVSSA DSTEKSASGA GTRNLPFQFC LRQALRMKAA GILTLIGCLV TGAESKIYTR CKLAKIFSRA GLDNYWGFSL GNWICMAYYE SGYNTTAQTV LDDGSIDYGI FQINSFAWCR RGKLKENNHC HVACSALITD DLTDAIICAR KIVKETQGMN YWQGWKKHCE GRDLSEWKKG CEVS; Positive Samples: Rat testis.

The lysozyme family of proteins has an important role in host defense and is associated with the monocyte-macrophage system and enhances the activity of immunoagents. The mammalian lysozyme gene family is much larger than previously appreciated and consists of at least eight distinct genes scattered around the genome. C-type lysozymes are specifically involved catalyzing the hydrolysis of Beta-1,4 glycosidic bonds of the peptidoglycan of bacterial cell walls. As a homolog of human C-type lysozyme, LYZL1 is an intra-acrosomal, nonbacteriolytic, c lysozyme-like protein recently isolated from human spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYZL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Lyzl1
Gene Accession No. Q6UWQ5
Gene Alias 1700038F02Rik; bA534G20.1; KAAG648; LYC2; lysozyme D1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; LYZD1; Lyzl1; PRO1278; RGD1559869; UNQ648/PRO1278
Gene Symbols Lyzl1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human LYZL1 (NP_1159063).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364745, 84569
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.