Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYZL1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304498100UL
Description
LYZL1 Polyclonal specifically detects LYZL1 in Rat samples. It is validated for Western Blot.Specifications
LYZL1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
EC 3.2.1.17, KAAG648, LYC2bA534G20.1, lysozyme-like 1, lysozyme-like protein 1, MGC33408, PRO1278 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human LYZL1 (NP_115906.3). | |
100 μL | |
Lipid and Metabolism | |
84569 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction