Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYZL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144149
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144149 Supplier Invitrogen™ Supplier No. PA5144149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYZL2 Polyclonal Antibody for IHC (P), ELISA

LYZL2, also known as Lysozyme-like 2, is a gene that encodes a protein belonging to the lysozyme family. Lysozymes are enzymes that play a role in the innate immune system by breaking down bacterial cell walls. LYZL2 shares structural similarities with lysozymes but has a distinct enzymatic activity. The specific function of LYZL2 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. It is worth noting that lysozymes and lysozyme-like proteins have been implicated in various biological processes, including antimicrobial defense, tissue remodeling, and reproductive functions. Therefore, understanding the specific functions and mechanisms of LYZL2 may provide valuable insights into these processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYZL2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 4.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LYZL2
Gene Accession No. Q7Z4W2
Gene Alias EC 3.2.1.17; Lysozyme-2; Lysozyme-like protein 2
Gene Symbols LYZL2
Host Species Rabbit
Immunogen Recombinant Human Lysozyme-like protein 2 protein (63-194AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119180
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.