Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LYZL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558547
Description
Immunogen sequence: YILGRCTVAK KLHDGGLDYF EGYSLENWVC LAYFESKFNP MAIYENTREG YTGFGLFQMR GSD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

Specifications
LYZL4 | |
Polyclonal | |
Unconjugated | |
LYZL4 | |
1810009N24Rik; AI504271; Lyc4; lysozyme A; lysozyme like 4; lysozyme like-4; lysozyme-4; lysozyme-like 4; lysozyme-like protein 4; Lyza; Lyzl4; RGD1308401 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
131375 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KX0 | |
LYZL4 | |
Recombinant protein corresponding to Human LYZL4. Recombinant protein control fragment (Product #RP-96387). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction