Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYZL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555250 Supplier Invitrogen™ Supplier No. PA555250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CNDYKSYSEN LCHVDCQDLL NPNLLAGIHC AKRIVSGARG MNNWVEWRLH CSGRP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.

Lysozymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYZL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lyzl6
Gene Accession No. O75951
Gene Alias 1700023H08Rik; epididymis secretory sperm binding protein Li 6a; HEL-S-6a; LOC540048 protein; Lyc1; lysozyme b; lysozyme homolog; lysozyme like 6; lysozyme-like 6; lysozyme-like protein 1; lysozyme-like protein 6; LYZB; Lyzl6; PRO1485; RGD1306968; TKAL754; UNQ754; UNQ754/PRO1485
Gene Symbols Lyzl6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LYZL6. Recombinant protein control fragment (Product #RP-93348).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.