Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZIC Antibody (OTI3A7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272559IR
Description
LZIC Monoclonal specifically detects LZIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
LZIC | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LZIC | |
Recombinant protein fragment corresponding to amino acids 1-190 of human LZIC (NP_115744) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3A7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
leucine zipper and CTNNBIP1 domain containing, Leucine zipper and CTNNBIP1 domain-containing protein, Leucine zipper and ICAT homologous domain-containing protein, leucine zipper domain and ICAT homologous domain containing, MGC15436, protein LZIC | |
Mouse | |
Affinity Purified | |
Neuroscience | |
84328.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction