Learn More
Invitrogen™ LZIC Monoclonal Antibody (OTI1D5)

Description
LZIC Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | LZIC |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LZIC |
| Gene Accession No. | Q8WZA0 |
| Gene Alias | 1810030J04Rik; AW047580; EGK_00210; leucine zipper and CTNNBIP1 domain containing; leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; leucine zipper domain and ICAT homologous domain containing; LZIC; Protein LZIC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.