Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LZIC Monoclonal Antibody (OTI5E3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526234

Catalog No. PIMA526234


Only null left
Add to Cart

Description

Description

LZIC Monoclonal Antibody for Western Blot, IHC (P)

LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LZIC
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQN7, Q8K3C3, Q8WZA0
LZIC
Full length human recombinant protein of LZIC produced in E.coli.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5E3
Unconjugated
LZIC
1810030J04Rik; AW047580; EGK_00210; leucine zipper and CTNNBIP1 domain containing; leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; leucine zipper domain and ICAT homologous domain containing; LZIC; Protein LZIC
Mouse
Affinity chromatography
RUO
100855658, 366507, 69151, 84328
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.