Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LZIC Monoclonal Antibody (OTI5E3), TrueMAB™, OriGene
SDP

Catalog No. 501676328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6328 Supplier OriGene Supplier No. TA505941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.

Specifications

Antigen LZIC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LZIC
Gene Accession No. Q5PQN7, Q8K3C3, Q8WZA0
Gene Alias leucine zipper and CTNNBIP1 domain containing
Gene Symbols LZIC
Host Species Mouse
Immunogen Full length human recombit protein of human LZIC produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855658, 366507, 69151, 84328
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.