Learn More
LZIC Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI5E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505941

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.Specifications
LZIC | |
Monoclonal | |
Unconjugated | |
LZIC | |
leucine zipper and CTNNBIP1 domain containing | |
Mouse | |
Affinity Chromatography | |
RUO | |
100855658, 366507, 69151, 84328 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5PQN7, Q8K3C3, Q8WZA0 | |
LZIC | |
Full length human recombit protein of human LZIC produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.