Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LZIC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535917
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (95%), Rat (96%), Zebrafish (86%), Chicken (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.
Specifications
| LZIC | |
| Polyclonal | |
| Unconjugated | |
| LZIC | |
| 1810030J04Rik; AW047580; EGK_00210; leucine zipper and CTNNBIP1 domain containing; leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; leucine zipper domain and ICAT homologous domain containing; LZIC; Protein LZIC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84328 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WZA0 | |
| LZIC | |
| Recombinant protein encompassing a sequence within the center region of human LZIC. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction