Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZIC Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572014
Description
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.
Specifications
LZIC | |
Polyclonal | |
Unconjugated | |
LZIC | |
Protein LZIC; Leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; LZIC | |
Rabbit | |
Protein A | |
RUO | |
366507, 84328 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.13 mg/mL | |
PBS with 0.09% sodium azide | |
Q5PQN7, Q8WZA0 | |
LZIC | |
KLH conjugated synthetic peptide between 81-109 amino acids from the Central region of human LZIC | |
400 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction